5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:120013-39-0
:
Formula:C₁₇H₂₄ClNO₃
MDL Number:MFCD09033179
:325.8304 g/mol
Melting Pt:250°C
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together