
1,3-Dimethoxy-1,3-dimethylurea ≥97.0% (by GC)
TCIAD3990-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
:
:148.16 g/mol
:42°C
:Refrigerator
:1993
:3,III
:GHS02
:123707-26-6
:C₅H₁₂N₂O₃