Order Entry
Italy
ContactUsLinkComponent
6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium perchlorate ≥98.0% (by HPLC)
 
 
6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium perchlorate ≥98.0% (by HPLC)
  TCIAD3982-100MG
 :  TCI
 :  
 
 
6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium perchlorate ≥98.0% (by HPLC)
  TCIAD3982-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%

 

 

 :
 :327.77 g/mol
 :1479
 :5.1,III
 :GHS03
 :GHS05
 :1334529-08-6
 :C₁₄H₁₈ClN₃O₄