
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:528-43-8
:
Formula:C₁₈H₁₈O₂
MDL Number:MFCD00016658
Merck Index:14,05697
Boiling Pt:180°C
:266.34 g/mol
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:99.0 to 103.0 deg-C
Specification Test Results
Frequently Bought Together