



Dimethylamine ∼10% ∼2 mol/L in THF
Supplier: TCI
Certificates
About this item
CAS Number:124-40-3
:
Formula:C₂H₇N
MDL Number:MFCD00008288
:45.08 g/mol
Merck Index:14,03228
Flash Pt:–36°C
Storage Temperature:Refrigerator
EINECS:204-697-4
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Concentration (Neutralization titration):10.0 to 12.0 w/w%
Specification Test Results
Frequently Bought Together