
4,4'-Dibromo-2,2'-bipyridyl ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:18511-71-2
:
Formula:C₁₀H₆Br₂N₂
MDL Number:MFCD00667763
:313.98 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:138.0 to 142.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together