Order Entry
Italy
ContactUsLinkComponent
Disodium prednisolone-21-phosphate ≥98.0% (by HPLC, titration analysis)
Health
Disodium prednisolone-21-phosphate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3918-5G
Supplier:  TCI
CAS Number:  
Health
Disodium prednisolone-21-phosphate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3918-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Specific rotation [a]20/D:
    +95.0 to +102.0 deg (C=1, pH 7.0 Buffer sol.) (calcd.on anh.substance)
  • Water:
    max. 8.0 %

Specification Test Results

About this item

 :
 :484.39 g/mol
Merck Index:14,07721
EINECS:204-722-9
 :GHS08
CAS Number:125-02-0
Formula:C₂₁H₂₉O₈P
MDL Number:MFCD00083682