Di-tert-butyl-L-(+)-tartrate ≥97.0% (by GC)
TCIAD3898-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:88.0 to 92.0 deg-C
- Specific rotation [a]20/D:+9.0 to +12.0 deg (C=1, Acetone)
:
:262.30 g/mol
:117384-45-9
:C₁₂H₂₂O₆
:MFCD00192000