
4,5-Dimethoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulphonate ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:866252-52-0
:
Formula:C₁₂H₁₇F₃O₅SSi
:358.40 g/mol
Storage Temperature:Freezer
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 90.0 %
Specification Test Results
Frequently Bought Together