Order Entry
Italy
ContactUsLinkComponent
 
 
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride ≥98.0% (by HPLC)
 :  TCI
 
 
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Drying loss:
    max. 1.0 %

 

Frequently Bought Together