
1,3-Dimesitylimidazol-2-ylidene ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:141556-42-5
:
Formula:C₂₁H₂₄N₂
MDL Number:MFCD09265338
:304.44 g/mol
Melting Pt:155°C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together