1,1-Dimethylsemicarbazide ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:22718-49-6
:
Formula:C₃H₉N₃O
:103.13 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:138.0 to 142.0 deg-C
Specification Test Results
Frequently Bought Together