2,9-Diphenyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3849-1G
Supplier: TCI
CAS Number:
Specifications
- Conf:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:186.0 to 190.0 deg-C
Specification Test Results
About this item
:
:332.41 g/mol
CAS Number:25677-69-4
Formula:C₂₄H₁₆N₂
MDL Number:MFCD00044208