(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:163169-29-7
:
Formula:C₂₈H₂₈FeNOP
MDL Number:MFCD15144869
:481.36 g/mol
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Chelometric Titration):min. 97.0 %
- Melting point:157.0 to 162.0 deg-C
Specification Test Results
Frequently Bought Together