Order Entry
Italy
ContactUsLinkComponent
Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) dimer ≥95.0% (by titrimetric analysis)
Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) dimer ≥95.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) dimer ≥95.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Chelometric Titration):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together