Order Entry
Italy
ContactUsLinkComponent
 
2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane ≥95.0% (by GC)
 :  TCI
 
2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    86.0 to 90.0 deg-C

 

Frequently Bought Together