
2,7-Dibromo-9,9'-spirobifluorene ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3781-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:474.1900 g/mol
:GHS07
CAS Number:171408-84-7
Formula:C₂₅H₁₄Br₂
MDL Number:MFCD08704218