
2,2'-Dithienyl disulphide ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:6911-51-9
:
Formula:C₈H₆S₄
MDL Number:MFCD00066333
:230.38 g/mol
Boiling Pt:132°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:56.0 to 60.0 deg-C
Specification Test Results
Frequently Bought Together