Di-1-naphthylmethanol ≥98.0% (by HPLC)
TCIAD3750-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:146.0 to 150.0 deg-C
:
:284.36 g/mol
:62784-66-1
:C₂₁H₁₆O
:MFCD11096937