3,5-Dimethyl-1-adamantanol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 1-Hydroxy-3,5-dimethyladamantane
CAS Number:707-37-9
:
Formula:C₁₂H₂₀O
MDL Number:MFCD00074775
:180.29 g/mol
Melting Pt:95°C
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:95.0 to 99.0 deg-C
Specification Test Results
Frequently Bought Together