Order Entry
Italy
ContactUsLinkComponent
 
Diethyl-trans-1,2-cyclohexanedicarboxylate ≥98.0% (by GC)
Catalog # TCIAD3684-5G
Supplier:  TCI
CAS Number:  
Diethyl-trans-1,2-cyclohexanedicarboxylate ≥98.0% (by GC)
Catalog # TCIAD3684-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :228.29 g/mol
Boiling Pt:104°C
CAS Number:17351-22-3
Formula:C₁₂H₂₀O₄