
Diphenylammonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:164411-06-7
:
Formula:C₁₃H₁₂F₃NO₃S
MDL Number:MFCD11042671
:319.30 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together