3,6-Dibromothieno[3,2-b]thiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:392662-65-6
:
Formula:C₆H₂Br₂S₂
MDL Number:MFCD10000958
:298.01 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:125.0 to 129.0 deg-C
Specification Test Results
Frequently Bought Together