
4,4'-Dithiodibutyric acid ≥99.0% (by GC, titration analysis)
Catalog # TCIAD3670-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:108.0 to 112.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:238.32 g/mol
EINECS:220-818-3
:GHS07
CAS Number:2906-60-7
Formula:C₈H₁₄O₄S₂
MDL Number:MFCD00004406