
4,6-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2434-56-2
:
Formula:C₄H₆N₄
MDL Number:MFCD00044572
:110.12 g/mol
Melting Pt:271°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together