
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1671-87-0
:
Formula:C₁₂H₈N₆
MDL Number:MFCD00121717
:236.24 g/mol
Melting Pt:225°C
:GHS07
Specification Test Results
- Appearance:Red to Dark red to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together