

1,3-Di(1-adamantyl)imidazolinium tetrafluoroborate ≥98.0% (by HPLC)
TCIAD3620-500MG
: TCI
:
- Pk:500 mg
- Seq:2725
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
:
:426.35 g/mol
:1759
:8,III
:GHS05
:GHS07
:1176202-63-3
:C₂₃H₃₅BF₄N₂