


Di-tert-butyl azodicarboxylate 20% in toluene
Catalog # TCIAD3544-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Concentration (Iodometric titration):20.0 to 23.0 w/w%
Specification Test Results
About this item
:
:230.26 g/mol
EINECS:212-796-9
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:870-50-8
Formula:C₁₀H₁₈N₂O₄
MDL Number:MFCD00015001