
(-)-Di-p-anisoyl-L-tartaric acid ≥97.0% (by HPLC, titration analysis)
TCIAD3491-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Optical purity (LC):min. 96.0 ee%
- Purity (Neutralization titration):min. 97.0 %
:
:418.35 g/mol
:GHS07
:50583-51-2
:C₂₀H₁₈O₁₀
:MFCD02682986