

6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd] ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:2010983-27-2
:
Formula:C₅₈H₇₀B₂F₈N₂O₂
:1000.82 g/mol
UN:1759
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together