DL-1,4-Dichloro-2,3-butanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2419-73-0
:
Formula:C₄H₈Cl₂O₂
:159.01 g/mol
Boiling Pt:152°C
EINECS:219-338-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:74.0 to 78.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together