
Dicyclopentanyl acrylate ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:7398-56-3
:
Formula:C₁₃H₁₈O₂
MDL Number:MFCD24663518
:206.29 g/mol
Flash Pt:132°C
:Stabilised with 4-methoxyphenol (MEHQ)
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomers)
Specification Test Results
Frequently Bought Together