4,4'-Dibromo-4''-phenyltriphenylamine ≥97.0% (by HPLC)
Catalog # TCIAD3303-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:136.0 to 140.0 deg-C
Specification Test Results
About this item
:
:479.22 g/mol
CAS Number:884530-69-2
Formula:C₂₄H₁₇Br₂N
MDL Number:MFCD08276360