9,9-Di-n-octylfluorene ≥97.0% (by HPLC)
Catalog # TCIAD3285-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White or Colorless to Yellow to Orange powder to lump to clear liquid
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:390.66 g/mol
CAS Number:123863-99-0
Formula:C₂₉H₄₂