

O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98.0% (by HPLC)
: TCI
:125700-69-8
:
:C₁₂H₁₆BF₄N₅O₂
:MFCD00077730
:349.10 g/mol
:1759
:8,III
:GHS05
:GHS07
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Frequently Bought Together