Order Entry
Italy
ContactUsLinkComponent
2,4-Diamino-6-[2-(2-methyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
2,4-Diamino-6-[2-(2-methyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
  TCIAD3238-25G
 :  TCI
 :  
2,4-Diamino-6-[2-(2-methyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
  TCIAD3238-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    251.0 to 256.0 deg-C

 

 

 : 6-(2-(2-Methyl-1H-imidazol-1-yl)ethyl)-1,3,5-triazine-2,4-diamine
 :
 :219.25 g/mol
 :254-074-6
 :GHS07
 :38668-46-1
 :C₉H₁₃N₇
 :MFCD00047350