
2,4-Diethylglutaric acid (DL- and meso- mixture) ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:27899-21-4
:
Formula:C₉H₁₆O₄
MDL Number:MFCD04117827
:188.22 g/mol
Melting Pt:78°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together