
2-(Diethylamino)ethyl bromide hydrobromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1069-72-3
:
Formula:C₆H₁₄BrN
MDL Number:MFCD00040376
:261.00 g/mol
Melting Pt:208°C
EINECS:213-962-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together