
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3207-200MG
Supplier: TCI
CAS Number:
Specifications
- Conf:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:240.0 to 244.0 deg-C
Specification Test Results
About this item
:
:284.362 g/mol
:GHS07
CAS Number:18741-85-0
Formula:C₂₀H₁₆N₂
MDL Number:MFCD00145204