1,5-Dibromoanthraquinone ≥93.0% (by HPLC)
Catalog # TCIAD3181-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 93.0 area%
Specification Test Results
About this item
:
:366.01 g/mol
Melting Pt:292°C
CAS Number:602-77-7
Formula:C₁₄H₆Br₂O₂