Order Entry
Italy
ContactUsLinkComponent
2,5-Diphenylthiophene ≥98.0% (by GC)
2,5-Diphenylthiophene ≥98.0% (by GC)
Catalog # TCIAD3177-100MG
Supplier:  TCI
CAS Number:  
2,5-Diphenylthiophene ≥98.0% (by GC)
Catalog # TCIAD3177-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    151.0 to 155.0 deg-C

Specification Test Results

About this item

 :
 :236.33 g/mol
CAS Number:1445-78-9
Formula:C₁₆H₁₂S
MDL Number:MFCD00626285