Order Entry
Italy
ContactUsLinkComponent
2,7-Dibromopyrene ≥97.0% (by GC)
Irritant
2,7-Dibromopyrene ≥97.0% (by GC)
Catalog # TCIAD3169-1G
Supplier:  TCI
CAS Number:  
Irritant
2,7-Dibromopyrene ≥97.0% (by GC)
Catalog # TCIAD3169-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :360.05 g/mol
Melting Pt:321°C
 :GHS07
CAS Number:102587-98-4
Formula:C₁₆H₈Br₂
MDL Number:MFCD09909862