Di-n-octyl sulphone ≥98.0% (by GC)
TCIAD3161-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:76.0 to 80.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:290.51 g/mol
:78°C
:7726-20-7
:C₁₆H₃₄O₂S
:MFCD00039760