4,5-Dibromo-N-methoxy-N-methyl-2-furancarboxamide ≥96.0% (by GC, titration analysis)
TCIAD2973-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Argentmetric Titration):min. 96.0 %
:
:312.95 g/mol
:502639-02-3
:C₇H₇Br₂NO₃