
4-(Diethylamino)benzaldehyde diphenylhydrazone ≥98.0% (by HPLC, total nitrogen)
TCIAD2952-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Light yellow to Yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:92.0 to 96.0 deg-C
- Solubility in hot Methanol:very faint turbidity
:
:343.47 g/mol
:269-181-3
:GHS07
:68189-23-1
:C₂₃H₂₅N₃
:MFCD00143309