
trans-N1,N2-Dimethylcyclohexane-1,2-diamine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:67579-81-1
:
Formula:C₈H₁₈N₂
MDL Number:MFCD03001702
:142.24 g/mol
Boiling Pt:83°C
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together