
2,8-Dichlorocyclooctanone ≥98.0% (by GC)
Catalog # TCIAD2910-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:195.08 g/mol
Boiling Pt:110°C
Melting Pt:43°C
:GHS07
Storage Temperature:Refrigerator
CAS Number:108249-93-0
Formula:C₈H₁₂Cl₂O
MDL Number:MFCD06797089