Order Entry
Italy
ContactUsLinkComponent
4,4'-Difluorochalcone ≥97.0% (by GC)
Irritant
4,4'-Difluorochalcone ≥97.0% (by GC)
Catalog # TCIAD2884-5G
Supplier:  TCI
CAS Number:  
Irritant
4,4'-Difluorochalcone ≥97.0% (by GC)
Catalog # TCIAD2884-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    117.0 to 121.0 deg-C

Specification Test Results

About this item

 :
 :244.24 g/mol
 :GHS07
CAS Number:2805-56-3
Formula:C₁₅H₁₀F₂O
MDL Number:MFCD00219906