2,2-Di-n-octyl-1,3-propanediol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106868-09-1
:
Formula:C₁₉H₄₀O₂
MDL Number:MFCD03844790
:300.53 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together