Order Entry
Italy
ContactUsLinkComponent
(S)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
(S)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
(S)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    +15.0 to +20.0 deg (C=0.5, EtOH (95%))

Specification Test Results

Frequently Bought Together